Skip to Content
Merck
vibrant-m

Cidofovir Related Compound A Hydrochloride

vibrant-m
Synonym(s):
1-[(S)-3-Hydroxy-2-(O-ethylphosphonomethoxy)propyl]cytosine
Empirical Formula (Hill Notation):
C10H18N3O6P
CAS Number:
Molecular Weight:
307.24

Products

Brand
Formula Weight
Agency
Quality Segment
Application
Package Size
Special Grade