Skip to Content
Merck
Chrysene BCR®, certified reference material

Chrysene

Chrysene BCR®, certified reference material
Synonym(s):
Chrysene
Empirical Formula (Hill Notation):
C18H12
CAS Number:
Molecular Weight:
228.29
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique