![Chlorzoxazone Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material, certified reference material, pharmaceutical secondary standard, pkg of 100 mg](/deepweb/assets/sigmaaldrich/product/structures/183/593/98cb97a5-6078-479e-b1e6-f0f038c5d228/640/98cb97a5-6078-479e-b1e6-f0f038c5d228.png)
Chlorzoxazone Related Compound A
![Chlorzoxazone Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material, certified reference material, pharmaceutical secondary standard, pkg of 100 mg](/deepweb/assets/sigmaaldrich/product/structures/183/593/98cb97a5-6078-479e-b1e6-f0f038c5d228/640/98cb97a5-6078-479e-b1e6-f0f038c5d228.png)
Synonym(s):
2-Amino-4-chlorophenol, 5-Chloro-2-hydroxyaniline
Linear Formula:
H2NC6H3(Cl)OH
CAS Number:
Molecular Weight:
143.57
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Agency
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1