
Chlorthalidone

Synonym(s):
2-Chloro-5(1-hydroxy-3-oxo-1- isoindolinyl) benzenesulfonamide, 2-Chloro-5-(2,3-dihydro-1-hydroxy-3-oxo-1H-isoindol-1-yl)benzenesulfonamide, Chlortalidone
Empirical Formula (Hill Notation):
C14H11ClN2O4S
CAS Number:
Molecular Weight:
338.77
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4