
Chloroplatinic acid solution

Synonym(s):
Hexachloroplatinic acid solution, Hydrogen hexachloroplatinate(IV) solution, Platinic chloride solution, Platinum(IV) chloride solution
Linear Formula:
H2PtCl6
CAS Number:
Molecular Weight:
409.81
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Reagent Type
Core
Showing 1-1 of 1