Cetyl palmitate 95
Synonym(s):
Palmityl palmitate, Cetyl palmitate, Hexadecyl hexadecanoate, Palmitic acid palmityl ester
Linear Formula:
CH3(CH2)14CO2(CH2)15CH3
CAS Number:
Molecular Weight:
480.85
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing