Skip to Content
Merck
Cerium(IV) sulfate tetrahydrate puriss. p.a., ≥98%

Cerium(IV) sulfate tetrahydrate

Cerium(IV) sulfate tetrahydrate puriss. p.a., ≥98%
Synonym(s):
Ceric sulfate tetrahydrate, Cerium(IV) sulfate tetrahydrate
Linear Formula:
Ce(SO4)2 · 4 H2O
CAS Number:
Molecular Weight:
404.30

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Reagent Type
Core
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing