Skip to Content
Merck
Carnosic acid primary reference standard

Carnosic Acid

Carnosic acid primary reference standard
Synonym(s):
Carnosic acid, (4aR,10aS)-5,6-Dihydroxy-7-isopropyl-1,1-dimethyl-1,3,4,9,10,10a-hexahydro-2H-phenanthrene-4a-carboxylic acid, Salvin
Empirical Formula (Hill Notation):
C20H28O4
CAS Number:
Molecular Weight:
332.43

Products

Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade