Carbon nanotube, multi-walled
Synonym(s):
MWCNT, MWNT, Multiwall carbon nanotube
CAS Number:
Products
Brand
Melting Point (°C)
Physical Form
Purity
Chemical Composition
Quality Segment
Particle Size (µm)
Showing 1-8 of 8
Compare
Product Number
Description
Pricing