Skip to Content
Merck
Calcium phosphate monobasic monohydrate purum p.a., ≥80% (KT)

Calcium phosphate monobasic monohydrate

Calcium phosphate monobasic monohydrate purum p.a., ≥80% (KT)
Synonym(s):
Calcium bis(dihydrogenphosphate) monohydrate, Calcium dihydrogenphosphate monohydrate, Primary calcium phosphate monohydrate
Linear Formula:
Ca(H2PO4)2 · H2O
CAS Number:
Molecular Weight:
252.07
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing