Calcium Ionophore A23187 mixed calcium magnesium salt
Synonym(s):
Calcimycin, Calcium Ionophore mixed calcium magnesium salt
Empirical Formula (Hill Notation):
C29H37N3O6
Molecular Weight:
523.62
Products
Brand
Formula Weight
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing