Calcitonin salmon
Synonym(s):
Thyrocalcitonin, Calcitonin, Salmon
Empirical Formula (Hill Notation):
C145H240N44O48S2
CAS Number:
Molecular Weight:
3431.85
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Sterility
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing