![Butalbital-D5 solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/162/096/ea7480f4-6160-421f-95ea-ea06a0cb5511/640/ea7480f4-6160-421f-95ea-ea06a0cb5511.png)
Butalbital-D5 solution
![Butalbital-D5 solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/162/096/ea7480f4-6160-421f-95ea-ea06a0cb5511/640/ea7480f4-6160-421f-95ea-ea06a0cb5511.png)
Synonym(s):
Butalbital-D5 solution
Empirical Formula (Hill Notation):
C11D5H11N2O3
CAS Number:
Molecular Weight:
229.29
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2