Bromodichloromethane
Synonym(s):
Bromodichloromethane, Dichlorobromomethane
Linear Formula:
BrCHCl2
CAS Number:
Molecular Weight:
163.83
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing