Skip to Content
Merck
Brevetoxin 3 solution 20 μg/g in acetonitrile (nominal concentration), certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Brevetoxin 3 solution

Brevetoxin 3 solution 20 μg/g in acetonitrile (nominal concentration), certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
42-Deoxo-42-hydroxybrevetoxin B solution, PbTx-3
Empirical Formula (Hill Notation):
C50H72O14
CAS Number:
Molecular Weight:
897.10

Products

Brand
Formula Weight
Manufacturer
Format
Quality Segment
Reference Material Type
Special Grade