![Brevetoxin 1 solution 20 μg/g in acetonitrile (nominal concentration), certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/312/698/d88d8fef-216b-40da-a0e0-7a37dd37c67a/640/d88d8fef-216b-40da-a0e0-7a37dd37c67a.png)
Brevetoxin 1 solution
![Brevetoxin 1 solution 20 μg/g in acetonitrile (nominal concentration), certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/312/698/d88d8fef-216b-40da-a0e0-7a37dd37c67a/640/d88d8fef-216b-40da-a0e0-7a37dd37c67a.png)
Synonym(s):
Brevetoxin A solution
Empirical Formula (Hill Notation):
C49H70O13
CAS Number:
Molecular Weight:
867.07
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1