![Boc-Gln-OH 98%](/deepweb/assets/sigmaaldrich/product/structures/638/020/2a5be27b-eba6-4693-8d9c-fc83afe95648/640/2a5be27b-eba6-4693-8d9c-fc83afe95648.png)
Boc-Gln-OH
![Boc-Gln-OH 98%](/deepweb/assets/sigmaaldrich/product/structures/638/020/2a5be27b-eba6-4693-8d9c-fc83afe95648/640/2a5be27b-eba6-4693-8d9c-fc83afe95648.png)
Synonym(s):
Nα-(tert-Butoxycarbonyl)-L-glutamine, Boc-L-glutamine
Linear Formula:
NH2COCH2CH2CH(COOH)NHCOOC(CH3)3
Products
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing