Bisacodyl Related Compound E
Synonym(s):
2-[(4-Acetoxyphenyl)(pyridin-2-yl)methyl]phenyl acetate
Empirical Formula (Hill Notation):
C22H19NO4
CAS Number:
Molecular Weight:
361.39
Products
Brand
Formula Weight
Agency
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing