
Betamethasone Dipropionate

Synonym(s):
Betamethasone 17,21-dipropionate, 9α-Fluoro-16β-methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione 17,21-dipropionate
Empirical Formula (Hill Notation):
C28H37FO7
CAS Number:
Molecular Weight:
504.59
Products
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2