![Benzoylecgonine-d3 solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/360/166/7a83b053-fe62-4585-9e1d-cd9257efe889/640/7a83b053-fe62-4585-9e1d-cd9257efe889.png)
Benzoylecgonine-d3 solution
![Benzoylecgonine-d3 solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/360/166/7a83b053-fe62-4585-9e1d-cd9257efe889/640/7a83b053-fe62-4585-9e1d-cd9257efe889.png)
Synonym(s):
Benzoylecgonine-N-methyl-d3 solution
Empirical Formula (Hill Notation):
C16D3H16NO4
CAS Number:
Molecular Weight:
292.34
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique