Skip to Content
Merck
Benz[a]anthracene BCR®, certified reference material

Benz[a]anthracene

Benz[a]anthracene BCR®, certified reference material
Synonym(s):
1,2-Benzanthracene, Tetraphene
Empirical Formula (Hill Notation):
C18H12
CAS Number:
Molecular Weight:
228.29
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Product Line
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
Reference Material Type
Special Grade
Technique