![Benoxacor certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/168/355/cf44cff9-04aa-4bae-9434-edcc80689098/640/cf44cff9-04aa-4bae-9434-edcc80689098.png)
Benoxacor
![Benoxacor certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/168/355/cf44cff9-04aa-4bae-9434-edcc80689098/640/cf44cff9-04aa-4bae-9434-edcc80689098.png)
Empirical Formula (Hill Notation):
C11H11Cl2NO2
CAS Number:
Molecular Weight:
260.12
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2