Azelastine Related Compound E
Synonym(s):
3-(4-Chlorobenzilidene)isobenzofuran-1(3H)-one
Empirical Formula (Hill Notation):
C15H9ClO2
CAS Number:
Molecular Weight:
256.68
Products
Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing