Azaperone
Synonym(s):
1-(4-Fluorophenyl)-4-(4-(2-pyridinyl)-1-piperazinyl)-1-butanone, 4′-Fluoro-4-[4-(2-pyridyl)-1-piperazinyl]butyrophenone
Empirical Formula (Hill Notation):
C19H22FN3O
CAS Number:
Molecular Weight:
327.40
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3