Skip to Content
Merck
Aurothioglucose hydrate ≥96% (titration)

Aurothioglucose hydrate

Aurothioglucose hydrate ≥96% (titration)
Synonym(s):
Gold thioglucose, Solganal, Solganol
Empirical Formula (Hill Notation):
C6H11AuO5S · xH2O
CAS Number:
Molecular Weight:
392.18 (anhydrous basis)
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing