![Atorvastatin Related Compound D Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/389/448/61dea3cb-8545-4a03-855a-13c8e732cfff/640/61dea3cb-8545-4a03-855a-13c8e732cfff.png)
Atorvastatin Related Compound D
![Atorvastatin Related Compound D Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/389/448/61dea3cb-8545-4a03-855a-13c8e732cfff/640/61dea3cb-8545-4a03-855a-13c8e732cfff.png)
Synonym(s):
3-[(4-fluorophenyl)carbonyl]-2-(2-methylpropanoyl)-N,3-diphenyloxirane-2-carboxamide, 3-(4-Fluorobenzoyl)-2-isobutyryl-N,3-diphenyloxirane-2-carboxamide, Atorvastatin Related Compound D
Empirical Formula (Hill Notation):
C26H22FNO4
CAS Number:
Molecular Weight:
431.46
Products
Brand
Formula Weight
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing