
Atorvastatin Calcium

Synonym(s):
Atorvastatin hemicalcium salt sesquihydrate
Empirical Formula (Hill Notation):
C66H68CaF2N4O10 · 3H2O
CAS Number:
Molecular Weight:
1209.39
Products
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2