![Astragaloside III phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/253/323/3fb5abf5-6a48-4317-86e1-4f7d90b0b3e1/640/3fb5abf5-6a48-4317-86e1-4f7d90b0b3e1.png)
Astragaloside III
![Astragaloside III phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/253/323/3fb5abf5-6a48-4317-86e1-4f7d90b0b3e1/640/3fb5abf5-6a48-4317-86e1-4f7d90b0b3e1.png)
Synonym(s):
(3β,6α,16β,20R,24S)-20,24-Epoxy-6,16,25-trihydroxy-9,19-cyclolanostan-3-yl 2-O-β-D-glucopyranosyl-β-D-xylopyranoside
Empirical Formula (Hill Notation):
C41H68O14
CAS Number:
Molecular Weight:
784.97
Products
Formula Weight
Manufacturer
Product Line
Format
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing