Skip to Content
Merck
Arachidonic acid sodium salt from Mortierella alpina ≥98.5% (GC)

Arachidonic acid sodium salt

Arachidonic acid sodium salt from Mortierella alpina ≥98.5% (GC)
Synonym(s):
5,8,11,14-Eicosatetraenoic acid, Eicosa-5Z,8Z,11Z,14Z-tetraenoic acid, Sodium arachidonate, cis,cis,cis,cis-5,8,11,14-Eicosatetraenoic acid sodium salt, 20:4
Empirical Formula (Hill Notation):
C20H31O2Na
CAS Number:
Molecular Weight:
326.45

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing