Aprotinin
Synonym(s):
BPTI, Bovine pancreatic trypsin inhibitor, Trasylol, Trypsin inhibitor (basic), Aprotinin from bovine lung
Empirical Formula (Hill Notation):
C284H432N84O79S7
CAS Number:
Molecular Weight:
6511.44
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Product Line
Purity
Tissue
Quality Segment
Package Size
Sterilization Method
Technique
Showing 1-8 of 8
Compare
Product Number
Description
Pricing