![Aphidicolin from Nigrospora sphaerica ≥98% (HPLC), powder](/deepweb/assets/sigmaaldrich/product/structures/154/853/ae64371e-4d3d-4320-973f-694facdc75a4/640/ae64371e-4d3d-4320-973f-694facdc75a4.png)
Aphidicolin from Nigrospora sphaerica
![Aphidicolin from Nigrospora sphaerica ≥98% (HPLC), powder](/deepweb/assets/sigmaaldrich/product/structures/154/853/ae64371e-4d3d-4320-973f-694facdc75a4/640/ae64371e-4d3d-4320-973f-694facdc75a4.png)
Synonym(s):
APC
Empirical Formula (Hill Notation):
C20H34O4
CAS Number:
Molecular Weight:
338.48
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Activity Spectrum
Quality Segment
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing