Anti-Rabbit IgG (whole molecule)−Gold antibody produced in goat
Products
Product Category
Brand
Biological Source
Clonality
Physical Form
Antibody Form
Research Area
Conjugate
Post Translational Modification
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing