
Anti-PKD2L2 antibody produced in rabbit

Synonym(s):
Anti-TRPP5, Anti-polycystic kidney disease 2-like 2
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing