Skip to Content
Merck
Anti-MAP Kinase Kinase (MEK, MAPKK) antibody produced in rabbit whole antiserum

Anti-MAP Kinase Kinase (MEK, MAPKK) antibody produced in rabbit

Anti-MAP Kinase Kinase (MEK, MAPKK) antibody produced in rabbit whole antiserum
Synonym(s):
Anti-MAPKK, Anti-MEK

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing