Skip to Content
Merck
Anti-Fibronectin antibody produced in rabbit affinity isolated antibody, buffered aqueous solution

Anti-Fibronectin Antibody

Anti-Fibronectin antibody produced in rabbit affinity isolated antibody, buffered aqueous solution
Synonym(s):
CIG, FINC, cold-insoluble globulin, fibronectin, fibronectin 1, Anti-CIG, Anti-FINC, Anti-GFND2, Anti-LETS, Anti-MSF, Anti-fibronectin 1, Anti-DKFZp686F10164, Anti-DKFZp686H0342, Anti-DKFZp686I1370, Anti-DKFZp686O13149, Anti-FN, Anti-Fibronectin 1

Products

Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Isotype
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Application
Post Translational Modification
Non-reactive Species
Species Reactivity
Technique
Showing 1-19 of 19
Compare
Product Number
Description
Pricing