Anti-ESR1 antibody produced in rabbit
Synonym(s):
Anti-ER antibody produced in rabbit, Anti-ER-α antibody produced in rabbit, Anti-Estradiol receptor antibody produced in rabbit, Anti-Estrogen receptor antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing