![Anti-β-Actin–FITC antibody, Mouse monoclonal clone AC-15, purified from hybridoma cell culture](/deepweb/assets/sigmaaldrich/product/images/157/022/93cd8d9c-08c7-4c75-81c1-f7cc1ad0bfa8/640/93cd8d9c-08c7-4c75-81c1-f7cc1ad0bfa8.jpg)
Anti-β-Actin–FITC antibody, Mouse monoclonal
![Anti-β-Actin–FITC antibody, Mouse monoclonal clone AC-15, purified from hybridoma cell culture](/deepweb/assets/sigmaaldrich/product/images/157/022/93cd8d9c-08c7-4c75-81c1-f7cc1ad0bfa8/640/93cd8d9c-08c7-4c75-81c1-f7cc1ad0bfa8.jpg)
Synonym(s):
Monoclonal Anti-β-Actin
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Non-reactive Species
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing