Skip to Content
Merck
Ammonia solution 25 % Suprapur®

Ammonia solution

Ammonia solution 25 % Suprapur®
Synonym(s):
Ammonium hydroxide solution, Ammonia aqueous, Ammonia water
Linear Formula:
NH4OH
CAS Number:
Molecular Weight:
35.05

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Agency
Format
Quality Segment
Application
Technique