
Amitraz

Synonym(s):
Amitraz, N′-(2,4-Dimethylphenyl)-N-{[(2,4-dimethylphenyl)imino]methyl}-N-methylmethanimidamide, NSC 324552
Empirical Formula (Hill Notation):
C19H23N3
CAS Number:
Molecular Weight:
293.41
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Special Grade
Showing 1-2 of 2