Skip to Content
BR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aluminum-magnesium-manganese-chromium alloy, Al96.45Mg2.6Mn0.8Cr0.15
Products
Manufacturer
Physical Form
ID (mm)
Length (mm)
OD (mm)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
GF63143119
Aluminum-magnesium-manganese-chromium alloy, Al96.45Mg2.6Mn0.8Cr0.15,
395mm tube, 2.1mm outside diameter, 0.25mm wall thickness, 1.6mm inside diameter, as drawn
Expand
View Pricing
GF45325717
Aluminum-magnesium-manganese-chromium alloy, Al96.45Mg2.6Mn0.8Cr0.15,
100mm tube, 2.1mm outside diameter, 0.25mm wall thickness, 1.6mm inside diameter, as drawn
Expand
View Pricing
GF10871658
Aluminum-magnesium-manganese-chromium alloy, Al96.45Mg2.6Mn0.8Cr0.15,
200mm tube, 2.1mm outside diameter, 0.25mm wall thickness, 1.6mm inside diameter, as drawn
Expand
View Pricing
GF14989258
Aluminum-magnesium-manganese-chromium alloy, Al96.45Mg2.6Mn0.8Cr0.15,
435mm tube, 2.1mm outside diameter, 0.25mm wall thickness, 1.6mm inside diameter, as drawn
Expand
View Pricing