Skip to Content
BR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aloin
Synonym(s):
1,8-Dihydroxy-10-(β-D-glucopyranosyl)-3-(hydroxymethyl)-9(10H)-anthracenone, 10-β-D-Glucopyranosyl-1,8-dihydroxy-3-(hydroxymethyl)-9(10H)-anthracenone, Aloin A, Barbaloin
Empirical Formula (Hill Notation):
C
21
H
22
O
9
CAS Number:
1415-73-2
Molecular Weight:
418.39
EC Number:
215-808-0
Products
Product Category
antimicrobials (1)
antioxidants (1)
antiprotozoal (1)
carbohydrates (1)
glycosides (1)
monosaccharides (1)
Show More
Brand
Sigma-Aldrich (2)
Biological Source
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Application
Optical Isomer
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
B6906
Aloin,
from
Aloe barbadensis
Miller leaves, ≥97%
Expand
View Pricing
A0451
Aloin,
from
Curacao aloe
, ~50%
Expand
View Pricing