![Aldicarb-(N-methyl-13C,d3 carbamoyl-13C) PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/353/433/f94e40f4-aeb3-47c9-b7e9-b1d462c660fc/640/f94e40f4-aeb3-47c9-b7e9-b1d462c660fc.png)
Aldicarb-(N-methyl-13C,d3 carbamoyl-13C)
![Aldicarb-(N-methyl-13C,d3 carbamoyl-13C) PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/353/433/f94e40f4-aeb3-47c9-b7e9-b1d462c660fc/640/f94e40f4-aeb3-47c9-b7e9-b1d462c660fc.png)
Synonym(s):
(1E)-2-Methyl-2-(methylthio)-O-[(methyl-13C-d3-amino)carbonyl-13C]oxime propanal
Empirical Formula (Hill Notation):
13C2C5D3H11N2O2S
CAS Number:
Molecular Weight:
195.27
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing