Alcian blue 8 GX (C.I.74240)
Synonym(s):
Alcian blue 8 GX (C.I.74240)
Empirical Formula (Hill Notation):
C56H68Cl4CuN16S4
CAS Number:
Molecular Weight:
1298.86
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing