Skip to Content
Merck
Acyclovir Pharmaceutical Secondary Standard; Certified Reference Material

Acyclovir

Acyclovir Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Acycloguanosine, 9-[(2-Hydroxyethoxy)methyl]guanine, Acyclovir
Empirical Formula (Hill Notation):
C8H11N5O3
CAS Number:
Molecular Weight:
225.20
EC Number:

Products

Brand
Formula Weight
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing