Skip to Content
Merck
Acetaldehyde dimethyl acetal 95%

Acetaldehyde dimethyl acetal

Acetaldehyde dimethyl acetal 95%
Synonym(s):
1,1-Dimethoxyethane, Dimethyl acetal, Acetaldehyde dimethyl acetal
Linear Formula:
CH3CH(OCH3)2
CAS Number:
Molecular Weight:
90.12

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing