Skip to Content
Merck
Acetaldehyde Pharmaceutical Secondary Standard; Certified Reference Material

Acetaldehyde

Acetaldehyde Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Ethanal, Acetaldehyde
Linear Formula:
CH3CHO
CAS Number:
Molecular Weight:
44.05

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Package Size
API Family
Reference Material Type
Special Grade
Technique