Skip to Content
Merck
7-Amino-4-(trifluoromethyl)coumarin ≥99%

7-Amino-4-(trifluoromethyl)coumarin

7-Amino-4-(trifluoromethyl)coumarin ≥99%
Synonym(s):
Coumarin 151
Empirical Formula (Hill Notation):
C10H6F3NO2
CAS Number:
Molecular Weight:
229.16
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing