![7-Acetyl-1,2,3,4,5,6,7,8-octahydro-1,1,6,7-tetramethylnaphthalene certified reference material, TraceCERT®, mixture of isomers, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/983/915/869e084a-aeb4-432f-9a2a-20b97dd91c43/640/869e084a-aeb4-432f-9a2a-20b97dd91c43.png)
7-Acetyl-1,2,3,4,5,6,7,8-octahydro-1,1,6,7-tetramethylnaphthalene
![7-Acetyl-1,2,3,4,5,6,7,8-octahydro-1,1,6,7-tetramethylnaphthalene certified reference material, TraceCERT®, mixture of isomers, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/983/915/869e084a-aeb4-432f-9a2a-20b97dd91c43/640/869e084a-aeb4-432f-9a2a-20b97dd91c43.png)
Synonym(s):
1-(1,2,3,4,5,6,7,8-Octahydro-2,3,8,8-tetramethyl-2-naphthalenyl)ethanone, 2-Acetyl-2,3,8,8-tetramethyloctalin, Iso E Super
Empirical Formula (Hill Notation):
C16H26O
CAS Number:
Molecular Weight:
234.38
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1