6-Tetrachloro-Fluorescein Phosphoramidite
Synonym(s):
6-(4, 7, 2′, 7′-Tetrachloro-3′,6′-dipivaloylfluoresceinyl-6-carboxamido)-hexyl-1-O-(2-cyanoethyl)-(N,N-diisopropyl)-phosphoramidite
Empirical Formula (Hill Notation):
C46H54N3O10Cl4P
Molecular Weight:
981.72
Products
Product Category
Brand
Color
Formula Weight
Purity
Showing 1-3 of 3