Skip to Content
BR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
(6S,7R)-6-[(BENZYLOXY)METHOXY]-7-(METHOXYMETHOXY)-5,5,11,11-TETRAMETHYLBICYCLO[6.2.1]UNDEC-1-ENE
Linear Formula:
C
25
H
38
O
4
CAS Number:
1217494-31-9
Molecular Weight:
402.579
Products
Formula Weight
Special Grade
Aldrich
CPR
(2)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
T295914
(6S,7R)-6-[(BENZYLOXY)METHOXY]-7-(METHOXYMETHOXY)-5,5,11,11-TETRAMETHYLBICYCLO[6.2.1]UNDEC-1-ENE,
Aldrich
CPR
Expand
View Pricing
T266205
(6S,7R)-6-[(BENZYLOXY)METHOXY]-7-(METHOXYMETHOXY)-5,5,11,11-TETRAMETHYLBICYCLO[6.2.1]UNDEC-1-ENE,
Aldrich
CPR
Expand
View Pricing